Learn More
Ace Glass, Inc. Tube, pressure, 15mL, 150psi, 25.4mm OD, 10.2cm long, complete with#15 front-seal plug
SDP
Catalog No. 509558493
Encompass_Preferred
15mL glass pressure tube with #15 Ace-Thred, rated at 150psig at as much as 120C. Includes #15 front-seal plug. Dimensions: 25.4mm O.D. x 10.2cm long.
Catalog No. 50-955-8493 Supplier Ace Glass, Inc. Supplier No. 864804
15mL glass pressure tube with #15 Ace-Thred, rated at 150psig at as much as 120C. Includes #15 front-seal plug. Dimensions: 25.4mm O.D. x 10.2cm long.