Learn More
Ace Glass, Inc. Tube, pressure, 15mL, 150psi, 25.4mm OD, 10.2cm long, complete with#15 back-seal plug
SDP
Catalog No. 509558492
Encompass_Preferred
15mL glass pressure tube with #15 Ace-Thred, rated at 150psig at as much as 120C. Includes #15 back-seal plug. Dimensions: 25.4mm O.D. x 10.2cm long.
Catalog No. 50-955-8492 Supplier Ace Glass, Inc. Supplier No. 864803
15mL glass pressure tube with #15 Ace-Thred, rated at 150psig at as much as 120C. Includes #15 back-seal plug. Dimensions: 25.4mm O.D. x 10.2cm long.