Learn More
Medchemexpress LLC 10-[3-(dimethylamino)-2-methylpropyl]phenothiazine-2-carbonitrile | 3546-03-0 | MFCD00868180 | 99.4% | C19H21N3S | 10MG
SDP
Catalog No. 5000260854
Encompass_Preferred
Cyamemazine is a phenothiazine-class neuroleptic used in research for its anxiolytic and antipsychotic properties. It functions as a serotonin receptor antagonist and is supplied as solid reference standards and DMSO solutions for pharmacology and biochemical assays. Chemical identifiers: CAS 3546-03-0; formula C19H21N3S; mol wt 323.46 g·mol⁻1.
  • Phenothiazine-class neuroleptic with anxiolytic and antipsychotic activity.
  • Antagonist at 5-HT3, 5-HT2A, and 5-HT2C receptors.
  • Available as solid standards and 10 mM DMSO solution for assay preparation.
  • High purity suitable for research use (99.4%).
  • Includes RP-HPLC documentation for quality verification.
Catalog No. 50-002-60854 Supplier Medchemexpress LLC Supplier No. HY1426410MG
Cyamemazine is a phenothiazine-class neuroleptic used in research for its anxiolytic and antipsychotic properties. It functions as a serotonin receptor antagonist and is supplied as solid reference standards and DMSO solutions for pharmacology and biochemical assays. Chemical identifiers: CAS 3546-03-0; formula C19H21N3S; mol wt 323.46 g·mol⁻1.
  • Phenothiazine-class neuroleptic with anxiolytic and antipsychotic activity.
  • Antagonist at 5-HT3, 5-HT2A, and 5-HT2C receptors.
  • Available as solid standards and 10 mM DMSO solution for assay preparation.
  • High purity suitable for research use (99.4%).
  • Includes RP-HPLC documentation for quality verification.