Learn More
Ace Glass, Inc. Connecting adapter with 10/30 top female and 14/20 bottom male standard taper joints.
SDP
Catalog No. 50976669
Encompass_Preferred
Connecting adapter with 10/30 top female and 14/20 bottom male standard taper joints.
Catalog No. 50-976-669 Supplier Ace Glass, Inc. Supplier No. 909208
Connecting adapter with 10/30 top female and 14/20 bottom male standard taper joints.