Learn More
Ace Glass, Inc. 10/30 straight-sided outer joint;
SDP
Catalog No. 50979339
Encompass_Preferred
10/30 straight-sided outer joint. O.D. of standard taper joint and tube are approximately the same size, allowing for ease in clamping.
Catalog No. 50-979-339 Supplier Ace Glass, Inc. Supplier No. 756713
10/30 straight-sided outer joint. O.D. of standard taper joint and tube are approximately the same size, allowing for ease in clamping.