Learn More
Integra Biosciences Corp 8 Channel VOYAGER, Adjustable Tip Spacing Pipette
SDP
Catalog No. NC1485418
Encompass
The VOYAGER pipette offers perfect balance, an ultra lightweight design, unsurpassed operational comfort. Easy to understand and intuitive to use, due to simple sliding with the finger over the touch wheel and full text menus. Using a VOYAGER, features a unique motorized tip spacing, enables parallel transfer of multiple samples between labware of different sizes and formats. The tip spacing can be changed by a simple push of a button.
Catalog No. NC1485418 Supplier Integra Biosciences Corp Supplier No. 4723
The VOYAGER pipette offers perfect balance, an ultra lightweight design, unsurpassed operational comfort. Easy to understand and intuitive to use, due to simple sliding with the finger over the touch wheel and full text menus. Using a VOYAGER, features a unique motorized tip spacing, enables parallel transfer of multiple samples between labware of different sizes and formats. The tip spacing can be changed by a simple push of a button.