Learn More
Ace Glass, Inc. Thermocouple well, 10/30 male ground joint, 5mm ID top, 1.8mm ID x 25mm below the grind immersion
SDP
Catalog No. NC9082213
Encompass_Preferred
Thermocouple well, 10/30 male ground joint, 5mm ID top, 1.8mm ID x 25mm below the grind immersion
Catalog No. NC9082213 Supplier Ace Glass, Inc. Supplier No. 909908
Thermocouple well, 10/30 male ground joint, 5mm ID top, 1.8mm ID x 25mm below the grind immersion