Learn More
Cole-Parmer Idex P-659 Threaded Luer Adapter, Natural PEEK, 0.050" Bore, Female Luer x Female 10-32 Coned; 1/EA
Catalog No. NC1797952 Shop All Cole-Parmer Products
Encompass
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
Catalog No. NC1797952 Supplier Cole-Parmer Supplier No. 0201415
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.