Learn More
Medchemexpress LLC 10-(4-(bis(2-acetoxyethyl)amino)phenyl)-BODIPY 505/515 | 886212-87-9 | 98.8% | 511.37 | C27H32BF2N3O4 | 10 MG
SDP
Catalog No. 5000297450
Encompass_Preferred
10-(4-(Bis(2-acetoxyethyl)amino)phenyl)-BODIPY 505/515 is a BODIPY-based small-molecule fluorescent probe designed for selective detection of Cu2+ ions. The compound is supplied as an orange to red solid with high chemical purity and is intended for fluorescence-based detection and imaging applications using a BODIPY 505/515 chromophore.
  • Selective fluorescent probe for Cu2+ ions.
  • BODIPY 505/515 fluorophore suitable for green fluorescence detection.
  • High supplied purity (≈98.8%).
  • Chemical formula C27H32BF2N3O4; molecular weight 511.37 g/mol.
  • Supplied as an orange to red solid in milligram pack sizes for laboratory use.
Catalog No. 50-002-97450 Supplier Medchemexpress LLC Supplier No. HYD155010MG
10-(4-(Bis(2-acetoxyethyl)amino)phenyl)-BODIPY 505/515 is a BODIPY-based small-molecule fluorescent probe designed for selective detection of Cu2+ ions. The compound is supplied as an orange to red solid with high chemical purity and is intended for fluorescence-based detection and imaging applications using a BODIPY 505/515 chromophore.
  • Selective fluorescent probe for Cu2+ ions.
  • BODIPY 505/515 fluorophore suitable for green fluorescence detection.
  • High supplied purity (≈98.8%).
  • Chemical formula C27H32BF2N3O4; molecular weight 511.37 g/mol.
  • Supplied as an orange to red solid in milligram pack sizes for laboratory use.