Learn More
NEST SCIENTIFIC INC 10.4ML 24-WELL DEEP WELL PLATE
Catalog No. NC2131470
Encompass_Preferred
Nest Scientific deep well plates are standardized to the SBS/ANSI footprint. This makes them ideal for high throuput worflows, as they are broadly compatible with automated instruments. Plates made of USP grade VI virgin polypropylene. Molded in an ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821).
This item is not returnable. View return policy
Catalog No. NC2131470 Supplier NEST SCIENTIFIC INC Supplier No. 504302
Nest Scientific deep well plates are standardized to the SBS/ANSI footprint. This makes them ideal for high throuput worflows, as they are broadly compatible with automated instruments. Plates made of USP grade VI virgin polypropylene. Molded in an ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821).