Learn More
NEST SCIENTIFIC USA 10.4 ml 24-well Deep Well Plate, V-Bottom, Square well, sterile, 50/cs
Catalog No. NC3650182
Encompass_Preferred
Nest Scientific deep well plates are standardized to the SBS/ANSI footprint. This makes them ideal for high throuput worflows, as they are broadly compatible with automated instruments. Plates made of USP grade VI virgin polypropylene. Molded in an ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821).
Catalog No. NC3650182 Supplier NEST SCIENTIFIC USA Supplier No. 504361
Add to Cart
Add to Cart
Nest Scientific deep well plates are standardized to the SBS/ANSI footprint. This makes them ideal for high throuput worflows, as they are broadly compatible with automated instruments. Plates made of USP grade VI virgin polypropylene. Molded in an ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821).