Learn More
Fine Science Tools 10 Fine Scissors Straight, 10.5cm
SDP
Catalog No. NC9551385
Encompass
The 10 Fine Scissors Straight 10.5cm are surgical scissors made by Sklar. They have a straight blade and are 10.5cm in length. The blades are made from stainless steel and have a fine tip. The handles have a tungsten carbide insert for durability and feature a contoured design for comfort during prolonged use. These scissors are suitable for delicate cutting tasks during surgical procedures. They can cut through soft tissue and fine sutures. The scissors are latex-free, corrosion resistant, and can be cleaned by autoclaving. They are sold individually.
Catalog No. NC9551385 Supplier Fine Science Tools Supplier No. 1406010
The 10 Fine Scissors Straight 10.5cm are surgical scissors made by Sklar. They have a straight blade and are 10.5cm in length. The blades are made from stainless steel and have a fine tip. The handles have a tungsten carbide insert for durability and feature a contoured design for comfort during prolonged use. These scissors are suitable for delicate cutting tasks during surgical procedures. They can cut through soft tissue and fine sutures. The scissors are latex-free, corrosion resistant, and can be cleaned by autoclaving. They are sold individually.