Learn More
Ace Glass, Inc. 10.5mm x 25cm chromatography column
SDP
Catalog No. 50972544
Encompass_Preferred
10.5mm x 25cm chromatography column with 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom.
Catalog No. 50-972-544 Supplier Ace Glass, Inc. Supplier No. 588810
10.5mm x 25cm chromatography column with 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom.