Learn More
Cole-Parmer General Tools 1478 Digital Stainless Steel Caliper, 0 to 8" with Fractions," or Milimeters UOM
Catalog No. NC0613085 Shop All Cole-Parmer Products
Encompass_Preferred
Direct inch/metric/fraction conversion; measure inside, outside, depth, step; friction thumb roller.
Catalog No. NC0613085 Supplier Cole-Parmer Supplier No. 9715224
Direct inch/metric/fraction conversion; measure inside, outside, depth, step; friction thumb roller.