Learn More
Rarecyte RareCyte AccuCyte CTC Sample Preparation Fluids
Catalog No. NC1374448
Encompass
Consists of three components that are used for processing ten blood samples for the detection of circulating tumor cells (CTCs). They include: Displacement Fluid (50mL), Isolation Fluid (2mL) and Transfer Fluid (8mL)
Catalog No. NC1374448 Supplier Rarecyte Supplier No. 241090002
Consists of three components that are used for processing ten blood samples for the detection of circulating tumor cells (CTCs). They include: Displacement Fluid (50mL), Isolation Fluid (2mL) and Transfer Fluid (8mL)