Learn More
Cayman Chemical 10-acetyl Docetaxel, Purity: ≥98%, CAS: 125354-16-7, 100 mg
SDP
Catalog No. 502951023
Encompass_Preferred
A derivative of paclitaxel and an inhibitor of microtubule depolymerization; inhibits calcium-induced depolymerization of calf brain MTP in a cell-free assay at 20 µM; cytotoxic to J774.2 cells (EC50 = 0.05 µM)
Catalog No. 50-295-1023 Supplier Cayman Chemical Supplier No. 34497100 MG
A derivative of paclitaxel and an inhibitor of microtubule depolymerization; inhibits calcium-induced depolymerization of calf brain MTP in a cell-free assay at 20 µM; cytotoxic to J774.2 cells (EC50 = 0.05 µM)