Learn More
NEW ENGLAND BIOGROUP 10 boxes of 50 LUNA-FX7 or LUNA-BX7 3-channel ultra low fluorescent counting slides, 500 slides/1500 counts. For LUNA-FX7 and LUNA-BX7.
SDP
Catalog No. NC3903957
Encompass
10 boxes of 50 LUNA-FX7 or LUNA-BX7 3-channel ultra low fluorescent counting slides, 500 slides/1500 counts. For LUNA-FX7 and LUNA-BX7.
Catalog No. NC3903957 Supplier NEW ENGLAND BIOGROUP Supplier No. L72022
10 boxes of 50 LUNA-FX7 or LUNA-BX7 3-channel ultra low fluorescent counting slides, 500 slides/1500 counts. For LUNA-FX7 and LUNA-BX7.