Learn More
Braintree Scientific 10 Boxes of 10 x 2mL 2% Tetra-EDTA Solution in 12mL syringe
SDP
Catalog No. 501956042
Encompass
T-FloLocTM 2% Tetra-EDTA Flush and Lock Solution PREMIUM catheter flushing and locking solution offering both patency and infection control. Provides Broad-Spectrum Activity - effective against gram positive & gram-negative bacteria and fungi associated with catheter-related infections Prevents and Eradicates Biofilm - eliminates protected microbial colonization of catheters effectively reducing the risk of catheter-related infection Effective Anticoagulant - promotes catheter patency; avoids systemic anticoagu­lation associated with heparin Favorable Safety Profile - non-toxic; no topical or systemic effects are known No Development of Resistance - does not induce development of drug-resistant bacte­rial strains NOTE: each syringe is sterile packaged, suitable for sterile field use **Sold for research purposes only and not for human use**
Catalog No. 50-195-6042 Supplier Braintree Scientific Supplier No. TFL120210
T-FloLocTM 2% Tetra-EDTA Flush and Lock Solution PREMIUM catheter flushing and locking solution offering both patency and infection control. Provides Broad-Spectrum Activity - effective against gram positive & gram-negative bacteria and fungi associated with catheter-related infections Prevents and Eradicates Biofilm - eliminates protected microbial colonization of catheters effectively reducing the risk of catheter-related infection Effective Anticoagulant - promotes catheter patency; avoids systemic anticoagu­lation associated with heparin Favorable Safety Profile - non-toxic; no topical or systemic effects are known No Development of Resistance - does not induce development of drug-resistant bacte­rial strains NOTE: each syringe is sterile packaged, suitable for sterile field use **Sold for research purposes only and not for human use**