Learn More
Medchemexpress LLC 10-Bromodecanoic acid | 50530-12-6 | 98.0% | 251.16 | 100 G
SDP
Catalog No. 5000454623
Encompass_Preferred
10-Bromodecanoic acid (Decanoic acid, 10-bromo-) is a linker that can be used in the synthesis of PROTACs, such as PROTAC KDM4 degrader-1 (HY-173135). It is for research use only and not sold to patients.
  • Protac linker
  • Can be used in the synthesis of protacs
Catalog No. 50-004-54623 Supplier Medchemexpress LLC Supplier No. HYW013381100G
10-Bromodecanoic acid (Decanoic acid, 10-bromo-) is a linker that can be used in the synthesis of PROTACs, such as PROTAC KDM4 degrader-1 (HY-173135). It is for research use only and not sold to patients.
  • Protac linker
  • Can be used in the synthesis of protacs