Learn More
Medchemexpress LLC 10-Bromodecanoic acid | 50530-12-6 | 98.0% | 251.16 | 50 G
SDP
Catalog No. 5000454176
Encompass_Preferred
10-Bromodecanoic acid (Decanoic acid, 10-bromo-) is a linker used in the synthesis of PROTACs.
  • Linker for PROTAC synthesis
Catalog No. 50-004-54176 Supplier Medchemexpress LLC Supplier No. HYW01338150G
10-Bromodecanoic acid (Decanoic acid, 10-bromo-) is a linker used in the synthesis of PROTACs.
  • Linker for PROTAC synthesis