Learn More
Medchemexpress LLC (10-Bromodecyl)phosphonic acid | 934985-98-5 | 99.4% | C10H22BrO3P | 25 MG
SDP
Catalog No. 5000352609
Encompass_Preferred
(10-Bromodecyl)phosphonic acid is an alkyl chain-based PROTAC linker used in the synthesis of PROTACs. PROTACs utilize two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase and the other targets the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system for selective protein degradation.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Exploits the intracellular ubiquitin-proteasome system
Catalog No. 50-003-52609 Supplier Medchemexpress LLC Supplier No. 25MGHY140330
(10-Bromodecyl)phosphonic acid is an alkyl chain-based PROTAC linker used in the synthesis of PROTACs. PROTACs utilize two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase and the other targets the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system for selective protein degradation.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Exploits the intracellular ubiquitin-proteasome system