Learn More
Medchemexpress LLC 10-bromodecylphosphonic acid | 934985-98-5 | 99.4% | C10H22BrO3P | 10MG
SDP
Catalog No. 5000260909
Encompass_Preferred
(10-Bromodecyl)phosphonic acid is a brominated alkyl phosphonic acid used as a linker building block in targeted protein degradation chemistry. It provides a 10-carbon chain bearing a phosphonic acid group and a terminal bromide for further functionalization, suitable for incorporation into PROTACs and related bifunctional molecules.
  • Alkyl chain-based linker suitable for PROTAC synthesis.
  • Phosphonic acid functional group enables stable conjugation and polarity control.
  • Terminal bromide allows nucleophilic substitution or coupling reactions.
  • High reported purity (99.44%) for reliable synthetic use.
  • Available in small milligram pack sizes for medicinal chemistry workflows.
  • Solid form with recommended cold storage for long-term stability.
Catalog No. 50-002-60909 Supplier Medchemexpress LLC Supplier No. HY14033010MG
(10-Bromodecyl)phosphonic acid is a brominated alkyl phosphonic acid used as a linker building block in targeted protein degradation chemistry. It provides a 10-carbon chain bearing a phosphonic acid group and a terminal bromide for further functionalization, suitable for incorporation into PROTACs and related bifunctional molecules.
  • Alkyl chain-based linker suitable for PROTAC synthesis.
  • Phosphonic acid functional group enables stable conjugation and polarity control.
  • Terminal bromide allows nucleophilic substitution or coupling reactions.
  • High reported purity (99.44%) for reliable synthetic use.
  • Available in small milligram pack sizes for medicinal chemistry workflows.
  • Solid form with recommended cold storage for long-term stability.