Learn More
Seracare Life Sciences Inc 10% BSA DILUENT/BLOCKING SOL
Supplier:  Seracare Life Sciences Inc 51400008
Encompass_Preferred
Convenient antibody diluent/blocking solution concentrate containing 10% bovine serum albumin in phosphate buffer. Compatible with most assay formats.
Catalog No. 50-674-07