Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Buffered Neutral Formalin (10%)

Catalog No. ES9710
Change view
Click to view available options
Quantity:
1 x 3.8 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES9710 1 x 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES9710 Supplier ENG Scientific Supplier No. 9710
Add to Cart
Add to Cart

Buffered Neutral Formalin (10%)

  • Buffered Neutral Formalin (10%)

Specifications

Chemical Name or Material Buffered Neutral Formalin
Packaging Bottle
Quantity 1 x 3.8 L
Concentration or Composition (by Analyte or Components) 10%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.