Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 10-camphorsulfonic acid ethyl ester | 108481-13-6 | >95.0% | 260.35 g/mol | C12H20O4S | 250 MG
SDP

Catalog No. 5000282502
Encompass_Preferred

10-Camphorsulfonic acid ethyl ester is the ethyl ester derivative of camphorsulfonic acid and is reported as an impurity of the antifungal voriconazole. It is provided for research and analytical use; molecular formula C12H20O4S and molecular weight 260.35 g/mol. Users should consult the certificate of analysis for detailed purity, storage, and handling information.

  • Ethyl ester derivative of camphorsulfonic acid.
  • Used as an impurity or analytical reference standard in pharmaceutical analysis.
  • Identified by CAS number 108481-13-6.
  • Molecular formula C12H20O4S and molecular weight 260.35 g/mol.
  • Storage and purity information provided in the certificate of analysis.

Catalog No. 50-002-82502 Supplier Medchemexpress LLC Supplier No. HYW329384250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

10-Camphorsulfonic acid ethyl ester is the ethyl ester derivative of camphorsulfonic acid and is reported as an impurity of the antifungal voriconazole. It is provided for research and analytical use; molecular formula C12H20O4S and molecular weight 260.35 g/mol. Users should consult the certificate of analysis for detailed purity, storage, and handling information.

  • Ethyl ester derivative of camphorsulfonic acid.
  • Used as an impurity or analytical reference standard in pharmaceutical analysis.
  • Identified by CAS number 108481-13-6.
  • Molecular formula C12H20O4S and molecular weight 260.35 g/mol.
  • Storage and purity information provided in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.