Learn More
Biotang Inc MS-H-S10 Analog Magnetic Hotplate Stirrer 10-Channel, s/steel plate, multi-position stirrer with 10 position, speed range 0-1100 rpm, Temperature range: Ambient to 120deg.C ,single stirring position interval 90mm max 0.4L for each position.
Catalog No. 507514916 Shop All BIOTANG Inc Products
Encompass_Preferred
MS-H-S10 Analog Magnetic Hotplate Stirrer 10-Channel, s/steel plate, multi-position stirrer with 10 position, speed range 0-1100 rpm, Temperature range: Ambient to 120deg.C ,single stirring position interval 90mm max 0.4L for each position.
Catalog No. 50-751-4916 Supplier Biotang Inc Supplier No. M10098
MS-H-S10 Analog Magnetic Hotplate Stirrer 10-Channel, s/steel plate, multi-position stirrer with 10 position, speed range 0-1100 rpm, Temperature range: Ambient to 120deg.C ,single stirring position interval 90mm max 0.4L for each position.