Learn More
Biotang Inc MS-M-S10 Analog Magnetic Stirrer 10-Channel, s/steel plate without Heating, multi-position stirrer with 10 position, speed range 0-1100 rpm, single stirring position interval 90mm max 0.4L for each position.
Catalog No. 507514915 Shop All BIOTANG Inc Products
Encompass_Preferred
MS-M-S10 Analog Magnetic Stirrer 10-Channel, s/steel plate without Heating, multi-position stirrer with 10 position, speed range 0-1100 rpm, single stirring position interval 90mm max 0.4L for each position.
Catalog No. 50-751-4915 Supplier Biotang Inc Supplier No. M10088
MS-M-S10 Analog Magnetic Stirrer 10-Channel, s/steel plate without Heating, multi-position stirrer with 10 position, speed range 0-1100 rpm, single stirring position interval 90mm max 0.4L for each position.