Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company 10cm x 1 cm Cell Adapter for DR 5000

Catalog No. 501998044 Shop All Hach Company Products
Encompass

Adapter for 10 cm x 1 cm rectangular cells

Catalog No. 50-199-8044 Supplier Hach Company Supplier No. LZY421
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adapter for 10 cm x 1 cm rectangular cells

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.