Learn More
Chemglass Life Sciences 10' Coiled Extension Cord, White, RTD
SDP
Catalog No. 501311227
Encompass_Preferred
10' Coiled Extension Cord, White, RTD. For use with CG-1978-P Series RTD sensors. An adapter having a female Lemo connector to the 2-blade mini plug is available to allow use of the “Style B” sensors with equipment having this connection, CG-3499-A-20
Catalog No. 50-131-1227 Supplier Chemglass Life Sciences Supplier No. CG394410
10' Coiled Extension Cord, White, RTD. For use with CG-1978-P Series RTD sensors. An adapter having a female Lemo connector to the 2-blade mini plug is available to allow use of the “Style B” sensors with equipment having this connection, CG-3499-A-20