Learn More
Apexbio Technology LLC 10-DAB (10-Deacetylbaccatin) 32981-86-5 200mg
SDP
Catalog No. 5000093126
Encompass_Preferred
10-Deacetylbaccatin is a diterpenoid intermediate used as a precursor in the synthesis of chemotherapeutic agents such as paclitaxel and docetaxel It serves as a starting framework for the chemical transformation into taxane-based antitumor compounds 10-Deacetylbaccatin exerts its value primarily through its utility in medicinal chemistry for developing and optimizing cancer therapeutics Based on these properties 10-Deacetylbaccatin holds research potential in the field of cancer drug development
Catalog No. 50-000-93126 Supplier Apexbio Technology LLC Supplier No. A4395200
10-Deacetylbaccatin is a diterpenoid intermediate used as a precursor in the synthesis of chemotherapeutic agents such as paclitaxel and docetaxel It serves as a starting framework for the chemical transformation into taxane-based antitumor compounds 10-Deacetylbaccatin exerts its value primarily through its utility in medicinal chemistry for developing and optimizing cancer therapeutics Based on these properties 10-Deacetylbaccatin holds research potential in the field of cancer drug development