Learn More
Selleck Chemical LLC 10-Deacetyl-7-xylosyltaxol S0563-25MG
SDP
Catalog No. 5000043665
Encompass_Preferred
10-Deacetyl-7-xylosyl paclitaxel (10-Deacetyl-7-xylosyltaxol 7-xylosyl-10-deacetylpaclitaxel) a derivative of paclitaxel and naturally occurring xyloside isolated from Taxus chinensis causes significant mitotic arrest in PC-3 cells followed by up-regulating expression of pro-apoptotic Bax and Bad protein as well as down-regulating expression of anti-apoptotic Bcl-2 and Bcl-XL which leads to a disturbance of the mitochondrial membrane permeability and to the activation of caspase-9
Catalog No. 50-000-43665 Supplier Selleck Chemical LLC Supplier No. S056325MG
10-Deacetyl-7-xylosyl paclitaxel (10-Deacetyl-7-xylosyltaxol 7-xylosyl-10-deacetylpaclitaxel) a derivative of paclitaxel and naturally occurring xyloside isolated from Taxus chinensis causes significant mitotic arrest in PC-3 cells followed by up-regulating expression of pro-apoptotic Bax and Bad protein as well as down-regulating expression of anti-apoptotic Bcl-2 and Bcl-XL which leads to a disturbance of the mitochondrial membrane permeability and to the activation of caspase-9