Learn More
Cayman Chemical 10-DeacetylbaccatIn III 10mg
SDP
Catalog No. 502987773
Encompass_Preferred
An inhibitor of microtubule assembly (IC50s = 23 and 0.9 µM for porcine brain and Physaram microtubules, respectively); a precursor in the semisynthetic synthesis of paclitaxel; inhibits growth of L. donovani intracellular amastigotes within J774 murine macrophages (IC50 = 70 nM)
Catalog No. 50-298-7773 Supplier Cayman Chemical Supplier No. 2226110 MG
An inhibitor of microtubule assembly (IC50s = 23 and 0.9 µM for porcine brain and Physaram microtubules, respectively); a precursor in the semisynthetic synthesis of paclitaxel; inhibits growth of L. donovani intracellular amastigotes within J774 murine macrophages (IC50 = 70 nM)