Learn More
Selleck Chemical LLC 10-Deacetylbaccatin-III S2409-25mg
SDP
Catalog No. 503112628
Encompass_Preferred
10-Deacetylbaccatin-III(10-DB III) is an antineoplastic agent and an anti-cancer intermediate
Catalog No. 50-311-2628 Supplier Selleck Chemical LLC Supplier No. S240925MG
10-Deacetylbaccatin-III(10-DB III) is an antineoplastic agent and an anti-cancer intermediate