Learn More
Medchemexpress LLC 10-Deacetylbaccatin III | 32981-86-5 | 99.79% | 544.59 | 200 MG
SDP
Catalog No. 5000391712
Encompass_Preferred
10-Deacetylbaccatin III (Standard) is the analytical standard of 10-Deacetylbaccatin III, intended for research and analytical applications. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Grade of analytical standard.
  • Reference standard supplied assay.
  • Commonly used in HPLC, GC, and MS research experiments.
Catalog No. 50-003-91712 Supplier Medchemexpress LLC Supplier No. HY16565200MG
10-Deacetylbaccatin III (Standard) is the analytical standard of 10-Deacetylbaccatin III, intended for research and analytical applications. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Grade of analytical standard.
  • Reference standard supplied assay.
  • Commonly used in HPLC, GC, and MS research experiments.