Learn More
Cayman Chemical 10-Deacetylbaccatin III, Purity: ≥98%, CAS: 32981-86-5, 50 mg
SDP
Catalog No. 502964279
Encompass_Preferred
An inhibitor of microtubule assembly (IC50s = 23 and 0.9 µM for porcine brain and Physaram microtubules, respectively); a precursor in the semisynthetic synthesis of paclitaxel; inhibits growth of L. donovani intracellular amastigotes within J774 murine macrophages (IC50 = 70 nM)
Catalog No. 50-296-4279 Supplier Cayman Chemical Supplier No. 2226150 MG
An inhibitor of microtubule assembly (IC50s = 23 and 0.9 µM for porcine brain and Physaram microtubules, respectively); a precursor in the semisynthetic synthesis of paclitaxel; inhibits growth of L. donovani intracellular amastigotes within J774 murine macrophages (IC50 = 70 nM)