Learn More
Enzo Life Sciences 10-DEBC hydrochloride (1mg). CAS: 201788-90-1
SDP
Catalog No. 502013415
Encompass_Preferred
Alternative name: 10-(4′-(N-diethylamino)butyl)-2-chlorophenoxazine . HCl, Akt inhibitor X. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO or methanol. Long Term Storage: -80°C.
Catalog No. 50-201-3415 Supplier Enzo Life Sciences Supplier No. ENZCHM1260001
Alternative name: 10-(4′-(N-diethylamino)butyl)-2-chlorophenoxazine . HCl, Akt inhibitor X. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO or methanol. Long Term Storage: -80°C.