Learn More
AdipoGen 10-Dodecylacridine Orange Br
SDP
Catalog No. 502561231
Encompass_Preferred
Chemical. CAS 41387-42-2. Formula C29H44BrN3. MW 514.58. Synthetic. Hydrophobic fluorescent probe. Selectively stains inner mitochondrial membranes, but in contrast to rhodamine-123 staining is not membrane potential-dependent. Dye binding to surfactant micellar surfaces and used for the determination of cationic surfactants.
Catalog No. 50-256-1231 Supplier AdipoGen Supplier No. CDXD0120M020
Chemical. CAS 41387-42-2. Formula C29H44BrN3. MW 514.58. Synthetic. Hydrophobic fluorescent probe. Selectively stains inner mitochondrial membranes, but in contrast to rhodamine-123 staining is not membrane potential-dependent. Dye binding to surfactant micellar surfaces and used for the determination of cationic surfactants.