Learn More
Cayman Chemical 10 E-PentadecenoIc AcId 10mg
SDP
Catalog No. 502986842
Encompass_Preferred
A 15-carbon monounsaturated fatty acid; inhibits IFN-γ-induced production of kynurenine in THP-1 cells by 14%; has been used in studies of alternative β-oxidation pathways
Catalog No. 50-298-6842 Supplier Cayman Chemical Supplier No. 2246710 MG
A 15-carbon monounsaturated fatty acid; inhibits IFN-γ-induced production of kynurenine in THP-1 cells by 14%; has been used in studies of alternative β-oxidation pathways