Learn More
Medchemexpress LLC 10(E)-pentadecenoic acid | 321744-58-5 | 99.9% | 1 MG
SDP
Catalog No. 5000234047
Encompass_Preferred
10(E)-pentadecenoic acid is a 15-carbon, long-chain monounsaturated fatty acid used as a research reagent. It has molecular formula C15H28O2, molecular weight 240.38 g/mol, and reported purity 99.9%. Supplied in milligram quantities for biochemical and lipid metabolism studies.
  • Long-chain monounsaturated fatty acid (15 carbons).
  • High reported purity suitable for analytical and biological research.
  • Available in small milligram quantities for assay development and standards.
  • Suitable for studies of lipid metabolism and β-oxidation pathways.
  • Provided with a product datasheet for analytical characterization.
Catalog No. 50-002-34047 Supplier Medchemexpress LLC Supplier No. HY1209961MG
Add to Cart
Add to Cart
10(E)-pentadecenoic acid is a 15-carbon, long-chain monounsaturated fatty acid used as a research reagent. It has molecular formula C15H28O2, molecular weight 240.38 g/mol, and reported purity 99.9%. Supplied in milligram quantities for biochemical and lipid metabolism studies.
  • Long-chain monounsaturated fatty acid (15 carbons).
  • High reported purity suitable for analytical and biological research.
  • Available in small milligram quantities for assay development and standards.
  • Suitable for studies of lipid metabolism and β-oxidation pathways.
  • Provided with a product datasheet for analytical characterization.