Learn More
Integra Biosciences Corp 1250 μl GripTips Sterile Filter, 10 Eco-Racks of 96 Tips
SDP
Catalog No. NC2340509
Encompass
The 1250 μL GripTips Sterile Filter are sterile, filtered pipette tips designed for the INTEGRA pipettes. They come in 10 eco-racks of 96 tips each, for a total of 960 tips. The tips have a maximum volume of 1250 μL and are made of medical grade virgin polypropylene. They feature a wide opening and a narrow tip end for accurate pipetting. The built-in hydrophobic filter prevents aerosols and liquids from reaching the pipette, ensuring sterility. These high quality tips offer reliable performance across a range of applications in the lab.
Catalog No. NC2340509 Supplier Integra Biosciences Corp Supplier No. 3745
The 1250 μL GripTips Sterile Filter are sterile, filtered pipette tips designed for the INTEGRA pipettes. They come in 10 eco-racks of 96 tips each, for a total of 960 tips. The tips have a maximum volume of 1250 μL and are made of medical grade virgin polypropylene. They feature a wide opening and a narrow tip end for accurate pipetting. The built-in hydrophobic filter prevents aerosols and liquids from reaching the pipette, ensuring sterility. These high quality tips offer reliable performance across a range of applications in the lab.