Learn More
Medchemexpress LLC 10-ethenyl-19-norprogesterone | 13258-85-0 | 99.7% | 326.47 g/mol | C22H30O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1315475MG
Encompass_Preferred
Org OD 02-0 (10-ethenyl-19-norprogesterone) is a steroidal membrane progesterone receptor α (mPRα) selective agonist used in research to probe mPRα-mediated signaling and progesterone-related biology. It activates MAPK signaling and inhibits pituitary prolactin secretion, and is provided as a solid for laboratory use.
  • mPRα selective agonist (reported IC50 ≈ 33.9 nM)
  • activates MAPK signaling
  • inhibits prolactin secretion in the pituitary
  • high purity: 99.7%
  • molecular formula C22H30O2; molecular weight 326.47 g/mol
  • available in small research quantities (example sizes: 1 mg, 5 mg, 10 mg, 50 mg)
Catalog No. 50-002-62178