Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azer Scientific 10% Formalin
SDP

Catalog No. 22225222
Change view
Click to view available options
Quantity:
55 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-225-222 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-225-222 Supplier Azer Scientific Supplier No. NBF55G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Neutral buffered formalin fixative is available in multiple size

  • Our 10% neutral buffered formalin fixative is available in multiple sizes including a 1 Gallon, 2.5 Gallon, 5 Gallon, 30 Gallon or 55 Gallon containers
  • 10% neutral buffered formalin is a general purpose fixative for tissues
  • No other dilution or addition of other agents is necessary
  • Penetrates quickly, but fixes slowly
  • Formulation is enhanced by buffering that optimizes histologic results by light microscopy and immunohistochemistry

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Formalin
Quantity 55 gal.
Solution Type Formalin
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.