Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab 10% Formalin, 55 Gal.
SDP

Catalog No. STL2860055
Change view
Click to view available options
Product Type:
Formalin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
STL2860055 Formalin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. STL2860055 Supplier StatLab Supplier No. 2860055
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Ready-to-use buffered formalin in bulk packaging.

  • Featuring a pH level of 7.0

Specifications

Capacity 55 gal.
Concentration Neutral Buffered 10% Formalin
For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Melting Point -134°F
Product Type Formalin
Quantity 55 gal.
Solution Type Formalin
Reagent Type Formalin Solution
Form Solution

Prefilled container

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.