Learn More
Medchemexpress LLC 10-formylfolic acid | 134-05-4 | MFCD22666228 | 94.5% | 469.41 g/mol | C20H19N7O7 | 10MG
SDP
Catalog No. 5000238494
Encompass_Preferred
10-Formylfolic acid is a folate derivative and biochemical research reagent that acts as a potent dihydrofolate reductase (DHFR) inhibitor. It is used as a probe in biochemical studies and has been reported as an early indicator in leukemia research. The compound is supplied in small research quantities and is accompanied by certificate of analysis and safety documentation.
  • Acts as a potent dihydrofolate reductase inhibitor.
  • Used as a biochemical probe and early indicator in leukemia research.
  • Available in multiple small pack sizes for research use.
  • Provided with certificate of analysis and safety data sheet for quality and handling.
  • Molecular formula C20H19N7O7 and molecular weight 469.41 g/mol.
Catalog No. 50-002-38494 Supplier Medchemexpress LLC Supplier No. HY12435010MG
Add to Cart
Add to Cart
10-Formylfolic acid is a folate derivative and biochemical research reagent that acts as a potent dihydrofolate reductase (DHFR) inhibitor. It is used as a probe in biochemical studies and has been reported as an early indicator in leukemia research. The compound is supplied in small research quantities and is accompanied by certificate of analysis and safety documentation.
  • Acts as a potent dihydrofolate reductase inhibitor.
  • Used as a biochemical probe and early indicator in leukemia research.
  • Available in multiple small pack sizes for research use.
  • Provided with certificate of analysis and safety data sheet for quality and handling.
  • Molecular formula C20H19N7O7 and molecular weight 469.41 g/mol.