Learn More
Medchemexpress LLC 10-formylfolic acid | 134-05-4 | MFCD28988728 | 94.5% | 469.41 g/mol | C20H19N7O7 | 5MG
SDP
Catalog No. 5000238581
Encompass_Preferred
10-Formylfolic acid is a folate derivative that acts as a potent inhibitor of dihydrofolate reductase. It is supplied as a research-grade analytical standard for biochemical and enzymatic studies and has been reported as an early indicator in leukemia research. The compound is intended for research use only and is not for clinical or human use.
  • Potent dihydrofolate reductase inhibitor suitable for enzyme assays.
  • Used in leukemia-related biochemical research and analytical studies.
  • Available in small, weighed quantities appropriate for analytical standards.
  • Characterized by chemical formula C20H19N7O7 and molecular weight 469.41 g/mol.
  • Provided with assay/purity data to support research reproducibility.
Catalog No. 50-002-38581 Supplier Medchemexpress LLC Supplier No. HY1243505MG
Add to Cart
Add to Cart
10-Formylfolic acid is a folate derivative that acts as a potent inhibitor of dihydrofolate reductase. It is supplied as a research-grade analytical standard for biochemical and enzymatic studies and has been reported as an early indicator in leukemia research. The compound is intended for research use only and is not for clinical or human use.
  • Potent dihydrofolate reductase inhibitor suitable for enzyme assays.
  • Used in leukemia-related biochemical research and analytical studies.
  • Available in small, weighed quantities appropriate for analytical standards.
  • Characterized by chemical formula C20H19N7O7 and molecular weight 469.41 g/mol.
  • Provided with assay/purity data to support research reproducibility.