Learn More
AirSea USA Salvage Drum
Supplier:  AirSea USA SD10
Encompass_Preferred
Salvage Drum. 10 Gallon, holds leaks and spills of chemicals and other hazards up to 7 gallons. For transport meeting US DOT regulation 173.3 ( c )
Catalog No. 50-153-4836