Learn More
Innovative Reseach Of America 10 Gauge Precision Trochar
SDP
Catalog No. NC9894995
Encompass
Stainless steel reusable precision trochar with regular medical point needle and rounded stylet protruding 1/8" from needle point for easy implantation of small pellets up to 3mm in diameter (usually 0.001 mg to to 5 mg pellet).
Catalog No. NC9894995 Supplier Innovative Reseach Of America Supplier No. MP182
Stainless steel reusable precision trochar with regular medical point needle and rounded stylet protruding 1/8" from needle point for easy implantation of small pellets up to 3mm in diameter (usually 0.001 mg to to 5 mg pellet).