Learn More
Medchemexpress LLC HY-N0448 5mg Medchemexpress, 10-Gingerol CAS:23513-15-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN04485MG
Encompass_Preferred
Medchemexpress, HY-N0448 5mg 10-Gingerol CAS:23513-15-7 10-Gingerol is a major pungent constituent in the ginger oleoresin from fresh rhizome, with anti-inflammatory, antioxidant and anti-proliferative activities. 10-Gingerol inhibits the proliferation of MDA-MB-231 tumor cell line with an IC50 of 12.1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0272