Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[10]-Gingerol, ≥94.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013630 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3630 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3630 Supplier MilliporeSigma Supelco Supplier No. 4263010MG
Add to Cart
Add to Cart
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 23513-15-7
Flash Point 166°C
Physical Form Neat
Linear Formula C21H34O4
MDL Number MFCD01682694
Synonym (S)- 5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-tetradecanone; 10-Gingerol
Type Analytical Standard
Quantity 10 mg
Formula Weight 350.49
Percent Purity ≥94.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.