Learn More
Electron Microscopy Sciences 10% Glutaraldehyde in Methanol 10 ML
SDP
Catalog No. 5024541
Encompass_Preferred
C₅H₈O₂
Rapid-Freezing on a polished metal surface, at the temp. of liquid helium or liquid nitrogen, followed by freeze-substitution in osmium-acetone, has been shown to be a very useful technique for capturing rapid cellular events and for preserving cell ultrastructure.
Catalog No. 50-245-41 Supplier Electron Microscopy Sciences Supplier No. 16532
C₅H₈O₂
Rapid-Freezing on a polished metal surface, at the temp. of liquid helium or liquid nitrogen, followed by freeze-substitution in osmium-acetone, has been shown to be a very useful technique for capturing rapid cellular events and for preserving cell ultrastructure.